Learn More
Neodymium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals
81.61 € - 272.36 €
Chemical Identifiers
CAS | 17522-69-9 |
---|---|
Molecular Formula | Cl3NdO12 |
Molecular Weight (g/mol) | 442.58 |
MDL Number | MFCD00198195 |
InChI Key | HBKMAYJLXKBOER-UHFFFAOYSA-K |
Synonym | neodymium 3+ perchlorate, neodymium perchlorate, neodymium 3+ triperchlorate, neodymium 3+ ion triperchlorate ion, neodymium iii perchlorate aqueous solution |
PubChem CID | 13783607 |
IUPAC Name | neodymium(3+);triperchlorate |
SMILES | [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Nd+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11430291
|
Thermo Scientific Alfa Aesar
040568.NB |
5 g |
81.61 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440291
|
Thermo Scientific Alfa Aesar
040568.ND |
25 g |
272.36 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neodymium(III) perchlorate used in the preparation of complexes of phosphine oxides. Two three-dimensional frameworks composed of doubly interpenetrating networks of the same topology but different stereoisomeric structures have been constructed from self-assembly of the chiral building block, Δ- or Λ-[Nd(ntb)2]3+.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17522-69-9 | |
442.58 | |
HBKMAYJLXKBOER-UHFFFAOYSA-K | |
13783607 | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Nd+3] |
Cl3NdO12 | |
MFCD00198195 | |
neodymium 3+ perchlorate, neodymium perchlorate, neodymium 3+ triperchlorate, neodymium 3+ ion triperchlorate ion, neodymium iii perchlorate aqueous solution | |
neodymium(3+);triperchlorate |
Specifications
17522-69-9 | |
Cl3NdO12 | |
UN3211 | |
Insoluble in water. | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Nd+3] | |
442.58 | |
550.68 (442.59 Anhydrous) | |
50% w/w aq. soln. | |
Neodymium(III) perchlorate |
Liquid | |
MFCD00198195 | |
neodymium 3+ perchlorate, neodymium perchlorate, neodymium 3+ triperchlorate, neodymium 3+ ion triperchlorate ion, neodymium iii perchlorate aqueous solution | |
HBKMAYJLXKBOER-UHFFFAOYSA-K | |
neodymium(3+);triperchlorate | |
13783607 | |
Reagent | |
Odorless |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H271-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
missing translation for 'einecsNumber' : 236-821-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.