Learn More
Neodymium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
90.45 € - 318.00 €
Chemical Identifiers
CAS | 34622-08-7 |
---|---|
Molecular Formula | C3F9NdO9S3 |
Molecular Weight (g/mol) | 591.432 |
MDL Number | MFCD00192343 |
InChI Key | WYRSPTDNOIZOGA-UHFFFAOYSA-K |
Synonym | neodymium iii trifluoromethanesulfonate, neodymium iii triflate, neodymium trifluoromethanesulfonate, trifluoromethanesulfonic acid neodymium iii salt, nd otf 3, neodymium triflate, acmc-209i8y, neodymium 3+ ion tritriflate, neodymium perfluoromethanesulfonate, neodymium 3+ ; trifluoromethanesulfonate |
PubChem CID | 2733937 |
IUPAC Name | neodymium(3+);trifluoromethanesulfonate |
SMILES | C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Nd+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11365667
|
Thermo Scientific Alfa Aesar
043393.09 |
10 g |
90.45 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11375667
|
Thermo Scientific Alfa Aesar
043393.18 |
50 g | N/A | ||||||
Beschreibung
Neodymium(III) trifluoromethanesulfonate is used in the Aldol reaction of silyl enol ethers with aldehydes and as a catalyst for dehydration of fructose, Diels-Alder reactions of cyclopentadiene and alkyl acrylates, α-amination reactions of pyrazolones, polymerization reactions of cyclosiloxanes, direct polycondensation of dicarboxylic acids and diols, regioselective reductive ring opening of benzylidene acetals, Paal-Knorr cyclocondensation and nitration of benzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
34622-08-7 | |
591.432 | |
WYRSPTDNOIZOGA-UHFFFAOYSA-K | |
2733937 | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Nd+3] |
C3F9NdO9S3 | |
MFCD00192343 | |
neodymium iii trifluoromethanesulfonate, neodymium iii triflate, neodymium trifluoromethanesulfonate, trifluoromethanesulfonic acid neodymium iii salt, nd otf 3, neodymium triflate, acmc-209i8y, neodymium 3+ ion tritriflate, neodymium perfluoromethanesulfonate, neodymium 3+ ; trifluoromethanesulfonate | |
neodymium(3+);trifluoromethanesulfonate |
Spezifikation
34622-08-7 | |
C3F9NdO9S3 | |
neodymium iii trifluoromethanesulfonate, neodymium iii triflate, neodymium trifluoromethanesulfonate, trifluoromethanesulfonic acid neodymium iii salt, nd otf 3, neodymium triflate, acmc-209i8y, neodymium 3+ ion tritriflate, neodymium perfluoromethanesulfonate, neodymium 3+ ; trifluoromethanesulfonate | |
WYRSPTDNOIZOGA-UHFFFAOYSA-K | |
neodymium(3+);trifluoromethanesulfonate | |
2733937 | |
98% | |
Neodymium (III) trifluoromethanesulfonate |
Powder | |
MFCD00192343 | |
Soluble in water. | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Nd+3] | |
591.432 | |
591.44 | |
1.7 g/mL |
Sicherheit und Handhabung
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.