missing translation for 'onlineSavingsMsg'
Learn More

Neohesperidin dihydrochalcone hydrate, 98+%, Thermo Scientific Chemicals

Product Code. 15465219
Click to view available options
Quantity:
1 g
5 g
25 g

Artificial sweetener derived from citrus

Neohesperidin dihydrochalcone hydrate, is a flavonoid sweetening agent with potent antioxidant activity. It is antioxidant agent. artificial sweetener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Neohesperidin dihydrochalcone hydrate, is a flavonoid sweetening agent with potent antioxidant activity. It is antioxidant agent. artificial sweetener.

Solubility
Soluble in water to 100 mM (with heating).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20702-77-6
Molecular Formula C28H36O15
Molecular Weight (g/mol) 612.58
MDL Number MFCD00017711
InChI Key ITVGXXMINPYUHD-UHFFFAOYSA-N
Synonym neohesperidin dihydrochalcone, nhdc, neohesperidin dc, neohesperidin dhc, unii-3x476d83qv, ccris 4848, neosperidin dihydrochalcone, neohesperidine dihydrochalcone, 1-4-2-o-6-deoxy-alpha-l-mannopyranosyl-beta-d-glucopyranosyl oxy-2,6-dihydroxyphenyl-3-3-hydroxy-4-methoxyphenyl propan-1-one, 1-4-2-o-6-deoxy-alpha-l-mannopyranosyl-beta-d-glucopyranosyl oxy-2,6-dihydroxyphenyl-3-3-hydroxy-4-methoxyphenyl
PubChem CID 30231
ChEBI CHEBI:83535
IUPAC Name 1-[4-[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-2,6-dihydroxyphenyl]-3-(3-hydroxy-4-methoxyphenyl)propan-1-one
SMILES COC1=C(O)C=C(CCC(=O)C2=C(O)C=C(OC3OC(CO)C(O)C(O)C3OC3OC(C)C(O)C(O)C3O)C=C2O)C=C1

Specifications

Color Yellow
Quantity 25 g
Merck Index 14,6452
Solubility Information Soluble in water to 100 mM (with heating).
Formula Weight 612.58 (Anhydrous)
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Neohesperidin dihydrochalcone hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'