missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neon™ NxT Electroporation System 10-μL Kit with 1-Channel Tubes

The Neon NxT Electroporation System 10-μL Kit is designed specifically for use with the Neon NxT Electroporation System.

776.00 € - 2285.00 €

Specifications

Cell Type Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format Electroporation Pipette Tip
Transfection Technique Electroporation
For Use With (Equipment) Neon™ NxT Electroporation System
For Use With (Application) Transfection
View More Specs

Products 4
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
17829444
View Documents
Invitrogen™
N1025
25 x 2 Reactions This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
18075331
View Documents
Invitrogen™
N1025
25 x 2 Reactions
776.00 €
Pack of 25
Please to purchase this item. Need a web account? Register with us today!
17839444
View Documents
Invitrogen™
N1096
96 x 2 Reactions This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
18035341
View Documents
Invitrogen™
N1096
96 x 2 Reactions
2285.00 €
Pack of 96
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Neon NxT Electroporation System 10-μL Kit with 1-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System (Cat. Nos. NEON1S, NEON1SK, NEON18S, NEON18SK). Use this kit for transfection volumes of 10 μL, containing 5 x 104–2 x 105 adherent cells or 1 x 105–5 x 105 suspension cells. Cells that have been transfected using the included 10-μL Neon NxT tips are ready to be washed and plated into a multi-well culture dish.

Specifications

Specifications

Stem Cells, Primary Cells, Hard-to-Transfect Cells
Electroporation
Transfection
10 μL
Electroporation Pipette Tip
Neon™ NxT Electroporation System
Pipette Tip
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.