Learn More
Invitrogen™ Neon™ NxT Electroporation System 8-Channel Pipette
The Neon NxT Electroporation System 8-Channel Pipette is designed specifically for use with the Neon NxT Electroporation System.
Brand:  Invitrogen™ NEON8P
Product Code. 30245209
  • 2854.00 € / Each
Explore more special offers
Description
Description
The Neon NxT Electroporation System 8-Channel Pipette is designed specifically for use with the Neon NxT Electroporation System (Cat. No. NEON18S). Each Neon NxT Electroporation System with 1-Channel and 8-Channel Pipettes (Cat. Nos. NEON18S and NEON18SK) comes with an 8-channel pipette. Those looking to upgrade their Neon NxT Electroporation System with the 1-Channel Pipette (Cat. Nos. NEON1S, NEON1SK) should pursue the Neon NxT Electroporation System 8-Channel Upgrade Package (Cat. Nos. NEON8 and NEON8U), which includes the Neon NxT Electroporation System 8-Channel Pipette, 8-channel pipette station, and 8-channel software activation. This pipette is offered separately for those needing an additional 8-channel pipette or replacement for their existing pipette.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transfection
Neon™ NxT Electroporation System
1 pipette
Neon™ NxT 10-μL and 100-μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2520624100089 Certificate TypeCertificate of Compliance Date27/01/2025 Catalog NumberNEON8P
Lot Number2520624100060 Certificate TypeCertificate of Compliance Date27/01/2025 Catalog NumberNEON8P
Lot Number2520624100083 Certificate TypeCertificate of Compliance Date27/01/2025 Catalog NumberNEON8P
Lot Number2520624090009 Certificate TypeCertificate of Compliance Date25/01/2025 Catalog NumberNEON8P
Lot Number2520624100037 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberNEON8P
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Neon™ NxT Electroporation System 8-Channel Pipette > Electroporation System 8-Channel Pipette