missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neon™ NxT Electroporation System Starter Pack

The Neon NxT Electroporation System Starter Kit with 1-Channel Pipette contains a Neon NxT Electroporation System with 1-channel pipette and pipette station as well as 192-reaction kits (10-μLL and 100-μLL sample sizes) to get you started in a convenient and cost-effective manner.

Brand:  Invitrogen™ NEON1SK

 View more versions of this product

Product Code. 17897694

  • 14140.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Flasks, Plates (6-well, 12-well, 24-well, and 96-well)f
Neon™ NxT Electroporation System
100/240 VAC
500–2500 V (Electroporation)
1–10 Pulses
Each
100–240 VAC, 50/60 Hz, 270 VA
Touchscreen
Transfection
Neon™ NxT Electroporation System Starter Pack
500–2500 V (Electroporation)
Color Touchscreen
1–100 ms
270 W
• Neon NxT electroporation device
• 1-Channel Pipette
• 1-Channel Pipette Station
• Power cord
• Quick reference guide
• Neon NxT Electroporation System 100 μL Kit (192 reactions); store buffers at 4°C
• Neon NxT Electroporation System 10 μL Kit (192 reactions); store buffers at 4°C
1-year Standard Warranty
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Neon™ NxT Electroporation System Starter Pack > NxT Electroporation System Starter Kit with 1-Channel Pipette

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.