Learn More
Neopentylamine, 97%, Thermo Scientific Chemicals
115.58 € - 326.00 €
Chemical Identifiers
CAS | 5813-64-9 |
---|---|
Molecular Formula | C5H13N |
Molecular Weight (g/mol) | 87.17 |
MDL Number | MFCD00008134 |
InChI Key | XDIAMRVROCPPBK-UHFFFAOYSA-N |
Synonym | neopentylamine, 2,2-dimethylpropylamine, 1-propanamine, 2,2-dimethyl, 2,2-dimethyl-1-propylamine, 1-amino-2,2-dimethylpropane, neo-c5h11nh2, 2,2-dimethyl-1-propylamin, neo-pentylamine, neopentyl amine, 2,2-dimethylpropanamine |
PubChem CID | 79882 |
IUPAC Name | 2,2-dimethylpropan-1-amine |
SMILES | CC(C)(C)CN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15254208
|
Thermo Scientific Acros
173690250 |
25 mL |
115.58 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15264208
|
Thermo Scientific Acros
173691000 |
100 mL |
326.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5813-64-9 | |
87.17 | |
XDIAMRVROCPPBK-UHFFFAOYSA-N | |
79882 | |
CC(C)(C)CN |
C5H13N | |
MFCD00008134 | |
neopentylamine, 2,2-dimethylpropylamine, 1-propanamine, 2,2-dimethyl, 2,2-dimethyl-1-propylamine, 1-amino-2,2-dimethylpropane, neo-c5h11nh2, 2,2-dimethyl-1-propylamin, neo-pentylamine, neopentyl amine, 2,2-dimethylpropanamine | |
2,2-dimethylpropan-1-amine |
Specifications
5813-64-9 | |
79.0°C to 82.0°C | |
Authentic | |
C5H13N | |
1.403 | |
04,188 | |
neopentylamine, 2,2-dimethylpropylamine, 1-propanamine, 2,2-dimethyl, 2,2-dimethyl-1-propylamine, 1-amino-2,2-dimethylpropane, neo-c5h11nh2, 2,2-dimethyl-1-propylamin, neo-pentylamine, neopentyl amine, 2,2-dimethylpropanamine | |
XDIAMRVROCPPBK-UHFFFAOYSA-N | |
2,2-dimethylpropan-1-amine | |
79882 | |
97% |
0.7400g/mL | |
−13°C | |
96% min. (GC) | |
(CH3)3CCH2NH2 | |
MFCD00008134 | |
0.74 | |
Solubility in water: soluble. | |
CC(C)(C)CN | |
87.17 | |
87.16 | |
Neopentylamine |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 227-378-1
- Neopentylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only