missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Neoprene Gloves
- One piece, seamless, 0.015 gauge
- With O-rings
- For use with Labconco Protector™ Glove Boxes, Precise™ Glove Boxes and XPert™ Weigh Boxes.
Specifications
Specifications
Color | Black |
Material | Neoprene |
Powdered | Powder Free |
Size | 8.5 |
Sterility | Non-sterile |
Packaging Type | 1 pair |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction