Learn More
Fisherbrand™ Neoprene Tubing
Used in flow lines for fluids and gases in a variety of applications.
Brand:  Fisherbrand™ FB50807
 View more versions of this product
Product Code. 11855502
  • 126.00 € / 10m
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Resistant to heat, ozone, petroleum oils and hydrocarbon solvents
  • Temperature range: -30°C to 100°C
Specifications
Specifications
6.5 x 10mm
Neoprene
10 m