missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Neotop™ 29-500 Series Neoprene Immersion Gloves

Ideal for use in cold conditions - neoprene maintains its superb elasticity even at low temperatures. Ansell Neotop™ 29-500 Series Neoprene Immersion Gloves protect against a wide range of acids, caustics, alcohols and many solvents.

Brand:  Ansell™ 29500100

44.94 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 11787583

  • 67.25 € / 12 pairs
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Ideal for use in cold conditions - neoprene maintains its superb elasticity even at low temperatures. Ansell Edmont™ Neotop™ 29-500 Series Neoprene Immersion Gloves protect against a wide range of acids, caustics, alcohols and many solvents.

  • Soft, comfortable and absorbant
  • Offers superior flexibility
  • Contains natural rubber
  • Much less tiring to the hands than other heavy-duty gloves
  • Flocklined in pure cotton
  • Applications include agriculture, chemical industries, oil and petrol refining, cleaning and maintenance
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rubber Latex
Black
Neoprene (Cuff), Cotton Flocking (Liner), Rubber Latex (Back)
10
0.75 mm
Straight
12 Pairs
29-500
Neoprene
Yes
Immersion Gloves
29-500
Black
Cotton Flocking
Acids, caustics, alcohols and many solvents
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers