missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Neotop™ 29-500 Series Neoprene Immersion Gloves

Product Code. 12455170
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 12455170

Brand: Ansell™ 29500090BP

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Ideal for use in cold conditions - neoprene maintains its superb elasticity even at low temperatures. Ansell Neotop™ 29-500 Series Neoprene Immersion Gloves protect against a wide range of acids, caustics, alcohols and many solvents.

Ideal for use in cold conditions - neoprene maintains its superb elasticity even at low temperatures. Ansell Edmont™ Neotop™ 29-500 Series Neoprene Immersion Gloves protect against a wide range of acids, caustics, alcohols and many solvents.

  • Soft, comfortable and absorbant
  • Offers superior flexibility
  • Contains natural rubber
  • Much less tiring to the hands than other heavy-duty gloves
  • Flocklined in pure cotton
  • Applications include agriculture, chemical industries, oil and petrol refining, cleaning and maintenance
TRUSTED_SUSTAINABILITY

Specifications

Back Material Rubber Latex
Cuff Material Neoprene
Applications Industrial
Color Black
Laboratory/Cleanroom Use Yes
Latex Free No
Material Neoprene (Cuff), Cotton Flocking (Liner), Rubber Latex (Back)
Product Type Immersion Gloves
Size 9
Thickness (English) 0.7 mil
Thickness (Metric) 0.75 mm
Glove Cuff Color Black
Glove Cuff Style Straight
Liner Material Cotton Flocking
Quantity 12 Pairs
Resistant To Acids, caustics, alcohols and many solvents
Series 29-500
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.