missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Edmont™ Scorpio™ 09-430 Neoprene Chemical Gloves
Shop All Ansell Products
Click to view available options
Size 10
Description
In environments where mechanical risks are combined with chemical exposure, the robust 09-430 series of neoprene gloves provides an optimal defense for the entire arm.
- Chemical and liquid protection gloves
- Top quality neoprene coating
- Cotton Jersey liner
- Protects against wide range of oils, acids, caustics, alcohols and many solvents
- Good resistance to all mechanical hazards
- Special Hi-Lo insulated gauntlet option with quilted cotton liner, for use in both hot and cold temperatures
- Permits intermittent handling in hot liquids up to 180°C and handling in cold temperatures down to -25°C
- Improves dexterity and reduces hand fatigue
- Certified FDA21 CFR 177-2600
Applications:
- Extruding, bending and punching shapes
- Fabricating metal parts, glass sheets
- Foundry operations
- Handling materials with sharp or rough edges
- Processing sheets, coils, tubes
- Unexpected leakages spills or other releases

Specifications
Specifications
Certifications/Compliance | EN 420:2003 + A1:2009 , EN 388:2006, and EN 374:2003 : 3121, ANSI, FDA 21 CFR 177.2600 |
Classification | Standard CATEGORY III |
Color | Black |
Exterior Finish | Smooth |
Hand | Specific |
Laboratory/Cleanroom Use | Yes |
Latex Free | Yes |
Material | Neoprene |
Product Type | Immersion Gloves |
Size | 10 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell Edmont™ Scorpio™ 09-430 Neoprene Chemical Gloves >
Spot an opportunity for improvement?Share a Content Correction