missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Edmont™ Neox™ 09-924 Series Black Neoprene Immersion Gloves
Click to view available options
Size 10
Description
- Top quality neoprene coating.
- Good resistance to all mechanical hazards.
- Comfortable anatomical glove shape which follows the contours of the hand at rest.
- Improves dexterity and reduces hand fatigue.
Specifications
Specifications
Back Material | Neoprene |
Certifications/Compliance | Standard CATEGORY III, EN 374, EN 388 : 3121, ANSI, FDA 21 CFR 177.2600 |
Cuff Material | Neoprene |
Coating Material | Neoprene |
Laboratory/Cleanroom Use | Yes |
Product Type | Immersion Gloves |
Style | 09-924 |
Temperature (Metric) Max | 180°C |
Glove Cuff Style | Gauntlet |
Lining | Cotton Jersey |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'