Learn More
Lovibond™ Tintometer™ Nessler Tube with Plungers
Used to assess product colour by comparison with representative series of fixed colour standards
Brand:  Lovibond™ 353080
Product Code. 13151671
  • 40.63 € / Pack of 2
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Anti-meniscus plungers
Specifications
Specifications
50 mL
113 mm
Nessleriser System 2150
2 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lovibond™ Tintometer™ Nessler Tube with Plungers > For Use With Nessleriser System 2150