missing translation for 'onlineSavingsMsg'
Learn More

Lovibond Tintometer™ Nessleriser Comparator

Lovibond Tintometer™ Nessleriser Comparator is designed for color matching of pale-colored samples where a long path length is desirable for accurate results.

Brand:  Lovibond Tintometer™ 172040

 View more versions of this product

Product Code. 13101671

  • 1160.00 € / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Lovibond Tintometer™ Nessleriser Comparator is designed for color matching of pale-colored samples where a long path length is desirable for accurate results.

  • Color comparator/colorimeter
  • Usess comparator disc standards
  • Determines chemical concentrations in solutions
  • Available options:
    • Tube length (250mm/113mm)
    • Lighting (lighting unit or ambient light)
    • DB 420 glass anti-meniscus plungers
Specifications

Specifications

Nessleriser 2250
Stand, daylight lighting unit and pair Nessler tubes, 250 mm with plungers
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers