missing translation for 'onlineSavingsMsg'
Learn More

Lovibond Tintometer™ Nessleriser Comparator

Lovibond Tintometer™ Nessleriser Comparator is designed for color matching of pale-colored samples where a long path length is desirable for accurate results.

Brand:  Lovibond Tintometer™ 172170

 View more versions of this product

Product Code. 13131671

  • 312.00 € / Each
Estimated Shipment: 23-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Lovibond Tintometer™ Nessleriser Comparator is designed for color matching of pale-colored samples where a long path length is desirable for accurate results.

  • Color comparator/colorimeter
  • Usess comparator disc standards
  • Determines chemical concentrations in solutions
  • Available options:
    • Tube length (250mm/113mm)
    • Lighting (lighting unit or ambient light)
    • DB 420 glass anti-meniscus plungers
Specifications

Specifications

Nessleriser 2250 Upgrade Kit
Pair Nessler tubes, 250 mm with plungers
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers