Learn More
NeuN Mouse anti-Human, Unconjugated, Clone: 3A4C1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66836-1-IG-150UL
This item is not returnable.
View return policy
Description
The NeuN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeuN. This antibody recognizes human, mouse, rat antigen. The NeuN antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
NeuN | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Immunohistochemistry, Immunofluorescence | |
3A4C1 | |
Unconjugated | |
150 μL | |
146713 | |
IgG1 |