Learn More
Hamilton™ Neuros™ 7000 Series Modified Microliter Syringes with Knurled Hub Needle
An industry first, the Neuros accurately dispenses 50nL to 100μL of liquid through an ultrafine needle protected by a custom-designed sleeve. The Neuros minimizes injection site damage by increasing needle rigidity and delivering a microvolume of liquid to an exact location.
Brand:  Hamilton™ 65457-01
 View more versions of this product
Product Code. 12646187
  • 504.00 € / Each
Explore more special offers
Description
Description
  • Needle rigidity improves insertion path accuracy
  • Minimal tissue damage reduces injection variability
  • Reduced sample loss saves money and materials
  • Fine gauge needle creates smaller injection sites
  • Compatibility with most infusion pumps and stereotaxic holders means an easy integration into existing processes
Specifications
Specifications
Autoclavable
32 G
0.004 in.
0.009 in.
0.103 mm
0.229 mm
Neuros™ adapter, two adjustable needle sleeves (one with a blind stop and one without)
1 in.
Microliter
239°F
50°F
3
Ea.
Blunt
Borosilicate Glass (Barrel), Tungsten Wire (Plunger)
0.004 in.
0.10 mm
0.308 in.
7.82 mm
Controlled animal injections
1 in.
0 to 20 mm
6000 psig (413.7 bar)
115°C
7000.5 KH
10°C
0.5 μL
Knurled Hub