missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurotensin Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00004922-B01P
This item is not returnable.
View return policy
Description
Neurotensin Polyclonal antibody specifically detects Neurotensin in Human samples. It is validated for Western Blot
Specifications
| Neurotensin | |
| Polyclonal | |
| PBS (pH 7.4) | |
| neurotensin, neurotensin/neuromedin N, NMN-125, NN, NT, NT/N, NTS1 | |
| NTS (NP_006174.1, 1 a.a. - 170 a.a.) full-length human protein. MMAGMKIQLVCMLLLAFSSWSLCSDSEEEMKALEADFLTNMHTSKISKAHVPSWKMTLLNVCSLVNNLNSPAEETGEVHEEELVARRKLPTALDGFSLEAMLTIYQLHKICHSRAFQHWELIQEDILDTGNDKNGKEEVIKRKIPYILKRQLYENKPRRPYILKRDSYYY | |
| 0.05 mg | |
| Stem Cell Markers | |
| 4922 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| NP_006174.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction