Learn More
Decon™ Neutracon
Decon Neutracon, Quantity: 5000mL, Amber, Format: Liquid, pH Range: 7.5, Viscosity: 55.0cps at 23 deg.C
Brand:  Decon™ NEU5
 View more versions of this product
Product Code. 10253710
  • 155.00 € / 5 L
Edge
Explore more special offers
Description
Description
Decon Neutracon, Quantity: 5000mL, Amber, Format: Liquid, pH Range: 7.5, Viscosity: 55.0cps at 23 deg.C
Biodegradable, phosphate, enzyme and chlorine bleach free, near-neutral liquid surface-active agent. Completely soluble in water and will not deposit calcium and magnesium or precipitate iron. Solutions are completely stable in air and do not absorb CO&superscript_2;. May be used in conjunction with water soluble solvents. A 13% solution is sufficient for the majority of cleaning applications. Heating the solution, or use in an ultrasonic cleaning bath will greatly reduce the cleaning time. Specifically formulated for cleaning/decontamination of non-ferrous metals, 'soft' silica glass, polymer surfaces and other materials which may be corroded, etched, degraded, discolored or weakened by acidic or alkaline cleaning agents.
Specifications
Specifications
Liquid
Amber
55.0cps at 23°C
7.5
Phosphate, Enzyme, Chlorine Bleach Free, Water
5000 mL