missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Neutral Red

Nuclear counterstain useful in pH range 6.8 to 8.0

35.58 € - 214.07 €

Chemical Identifiers

CAS 553-24-2
Molecular Formula C15H17ClN4
Molecular Weight (g/mol) 288.779
MDL Number MFCD00012651
InChI Key PGSADBUBUOPOJS-UHFFFAOYSA-N
Synonym Basic Red 5; C.I. 50040
PubChem CID 11105
ChEBI CHEBI:86370
IUPAC Name 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
SMILES CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15475319
View Documents
Thermo Scientific Chemicals
J63054.03
1 g
35.58 €
1g
In Stock
to see stock.
Add to basket
 
15485319
View Documents
Thermo Scientific Chemicals
J63054.06
5 g
64.32 €
5g
In Stock
to see stock.
Add to basket
 
15495319
View Documents
Thermo Scientific Chemicals
J63054.14
25 g
214.07 €
25g
In Stock
to see stock.
Add to basket
 
Description

Description

Nuclear counterstain useful in pH range 6.8 - 8.0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

553-24-2
288.779
PGSADBUBUOPOJS-UHFFFAOYSA-N
11105
8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
C15H17ClN4
MFCD00012651
Basic Red 5; C.I. 50040
CHEBI:86370
CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl

Specifications

290°C (decomposition)
C15H17ClN4
3918740
Basic Red 5; C.I. 50040
CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
288.779
CHEBI:86370
Black to Green
Neutral Red
553-24-2
MFCD00012651
14,6488
PGSADBUBUOPOJS-UHFFFAOYSA-N
8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
11105
288.78
Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 209-035-8

missing translation for 'rtecsNumber' : SG1400000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.