missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Neutral Red

Product Code. 10268473
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-24-2
Molecular Formula C15H17ClN4
Molecular Weight (g/mol) 288.779
MDL Number MFCD00012651
InChI Key PGSADBUBUOPOJS-UHFFFAOYSA-N
Synonym Basic Red 5, C.I. 50040, 3-Amino-7-dimethylamino-2-methylphenazine hydrochloride
PubChem CID 11105
ChEBI CHEBI:86370
IUPAC Name 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
SMILES CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl

Specifications

Melting Point 290.0°C
Absorption Ratio 1 to 1.12
Dye Content 50% min.
Beilstein 25, 401
Merck Index 15, 6573
Solubility Information Solubility in water: 50g/L. Other solubilities: 1.8% in abs. alcohol, 3.75% in cellosolve, 3.0% in ethylene glycol, practically insoluble in xylene
Infrared Spectrum Authentic
Lambda Max 539 to 544nm
Formula Weight 288.77
Grade Pure
Packaging Glass bottle
Color Green
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material Neutral Red, Pure
Show More Show Less
Product Identifier
  • Neutral Red

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.