missing translation for 'onlineSavingsMsg'
Learn More

Neutralization solution, Thermo Scientific Chemicals

33.80 € - 60.80 €

Specifications

Chemical Name or Material Neutralization solution
MDL Number MFCD00283950
Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15493059
View Documents
Thermo Scientific Alfa Aesar
J60275.AK
250 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15403069
View Documents
Thermo Scientific Alfa Aesar
J60275.AP
500 mL
60.80 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Neutralization solution
MFCD00283950
Liquid
Videos
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only