missing translation for 'onlineSavingsMsg'
Learn More

New Pig Haz-Mat Pads

Stop the spread of acids and bases.

Brand:  New Pig MAT310

Encompass_Preferred

 View more versions of this product

Product Code. 12419277

  • 74.74 € / Pack of 50
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Stop the spread of acids and bases.

  • Chemical-resistant — won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Excellent for quickly diking and stopping the spread of chemical spills
  • Absorb and retain most acids, bases and unknown liquids
  • Can be incinerated after use to reduce waste

Specifications

Specifications

Pink
0.833 L
Hazmat Absorbent Pad
20 L x 15 in. W (50.8 x 38 cm)
20 x 15 in. (50.8 x 38 cm)
50.8 cm
38 cm
3.6 kg
28.16 oz.
Polypropylene
Quickly diking and stopping the spread of chemical spills, absorbs acids, bases, corrosives
Heavyweight
20 in.
15 in.
8 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers