missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Newborn Bovine Serum
Used in cell culture to support the maturation of oocytes, to study the function of tubular epithelial cells, and to develop primary cultures of cells such as visceral adipocytes.
31.16 € - 121.90 €
Specifications
Color | Golden Brown |
---|---|
Content And Storage | -20°C |
Product Type | Newborn Calf Bovine Serum |
Sterilization Method | 0.2 μm filtration |
Source | Bovine Neonate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17347479
|
MP Biomedicals™
092912149 |
100 mL |
31.16 €
100mL |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11405025
|
MP Biomedicals™
092912154 |
500 mL |
121.90 €
500mL |
Estimated Shipment: 26-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Golden Brown | |
Newborn Calf Bovine Serum | |
Bovine Neonate | |
Cell culture and organ preservation applications | |
No | |
265 to 323mOsm/kg | |
4.4 to 7% |
-20°C | |
0.2 μm filtration | |
30EU/mL | |
≤27mg/dL | |
No | |
7.1 to 8.2 |