missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF 546, Tocris Bioscience™
Click to view available options
10mg
Description
P2Y11 agonist (pEC50 = 6.27). Exhibits selectivity for P2Y11 over a range of P2Y and P2X receptors. Also inhibits DNA-binding activity of HMGA2 (IC50 = 5.49 μM).
Specifications
CAS | 1006028-37-0 |
Molecular Formula | C47H44N6Na4O17P4 |
Quantity | 10 mg |
Synonym | d03pea, 4,4'-carbonylbis imino-3,1-phenylene-carbonylimino-3,1-4-methyl-phenylene carbonylimino-bis 1,3-xylene-alpha,alpha'-diphosphonic acid tetrasodium salt |
InChI Key | GSMUPMANKNAMAS-UHFFFAOYSA-J |
SMILES | CC1=C(C=C(C=C1)C(=O)NC2=C(C=C(C=C2)CP(=O)(O)[O-])CP(=O)(O)[O-])NC(=O)C3=CC(=CC=C3)NC(=O)NC4=CC=CC(=C4)C(=O)NC5=C(C=CC(=C5)C(=O)NC6=C(C=C(C=C6)CP(=O)(O)[O-])CP(=O)(O)[O-])C.[Na+].[Na+].[Na+].[Na+] |
IUPAC Name | tetrasodium;[2-[[3-[[3-[[3-[[5-[[2,4-bis[[hydroxy(oxido)phosphoryl]methyl]phenyl]carbamoyl]-2-methylphenyl]carbamoyl]phenyl]carbamoylamino]benzoyl]amino]-4-methylbenzoyl]amino]-5-[[hydroxy(oxido)phosphoryl]methyl]phenyl]methyl-hydroxyphosphinate |
Molecular Weight (g/mol) | 1180.748 |
PubChem CID | 90488895 |
Formula Weight | 1180.74 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'