Learn More
Tocris™ Tasquinimod
Tasquinimod is a high affinity negative allosteric modulator of HDAC4 (Kd = 10 to 30 nM).
Brand:  Tocris™ 7051/5
94.17 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 30000935
  • 120.00 € / 5mg
Explore more special offers
Description
Description
Tasquinimod is a high affinity negative allosteric modulator of HDAC4 (Kd = 10 - 30 nM). Binds the regulatory Zn2+ binding domain of HDAC4. Suppresses hypoxia-induced decrease in histone acetylation in human prostate cancer cells in vitro. Also binds S100A9. Antiangiogenic. Inhibits endothelial sprouting in vitro and growth of prostate tumor xenografts in nude mice. Orally bioavailable.
Specifications
Specifications
  • Store kit components at the temperatures indicated on the labels and insert until their expiration date
  • Store HeLa Whole Cell Extract at -80°C. Avoid repeated freeze-thaw cycles. Stable at -80°C for 6 months
1 Plate