Learn More
R&D Systems NHP XL CXCL2/GRO beta, Magnetic Luminex™ Performance Assay
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand:  R&D Systems LNHPXL276
Product Code. 18774544
  • 227.00 € / 1 set
Explore more special offers
Specifications
1 Kit
CXCL2/GRO beta/MIP-2/CINC-3
chemokine (C-X-C motif) ligand 2, CINC-2a, CINC3, CINC-3, C-X-C Motif Chemokine 2, GRO beta, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MGSA-beta, MIP2, MIP-2, MIP-2a, MIP2A, MIP2-alpha, SCYB2
Primate
2920
Store the unopened product at 2 - 8 °C. Do not use past expiration date.