Learn More
Cytiva NHS-Activated Sepharose™ 4 Fast Flow Affinity Media
Use for coupling amino-containing smaller proteins and peptides. Cytiva Life Sciences™ NHS-Activated Sepharose™ 4 Fast Flow Affinity Media yields coupled products that are very stable, especially at high pH.
Brand:  Cytiva 17-0906-01
Product Code. 10343240
  • 312.00 € / 25mL
Explore more special offers
Description
Description
  • Composed of cross-linked 4% agarose beads, NHS activated for coupling amino-containing proteins and peptides
  • NHS (N-hydroxysuccinimide) coupling forms chemically stable amide bond with ligands containing primary amino groups
  • Resin provides spacer arm to coupled ligand
  • Fast Flow matrix gives high productivity
  • Coupled product is very stable, especially at high pH
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 mL
For Coupling Amino-Containing Smaller Proteins and Peptides
NHS-Activated Sepharose 4 Fast Flow Media
45 to 165 μm
25 cm
4% Cross-Linked Agarose
100% Isopropanol
Stable in commonly used aqueous Buffers provided that the ligand withstand the pH or the chemical environment
2°C to 8°C
150 to 250 cm/Hoμr
9.84 in.
100 kPa
3 to 13
N-hydroxysuccinimide (NHS) Activated
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva NHS-Activated Sepharose™ 4 Fast Flow Affinity Media > Pack size: 25mL