missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NHS-Fluorescein (5/6-carboxyfluorescein succinimidyl ester), mixed isomer

Product Code. p-7184557 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
This item is not returnable. View return policy

Product Code. 10280534

Brand: Thermo Scientific™ 46409

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Activated derivatives of green-fluorescent fluorescein dye to label antibodies, proteins and other molecules for fluorescence imaging (ex 494; em 518).

Thermo Scientific™ FITC and Fluorescein Labeling Reagents and Kits are high-performance derivatives of fluorescein dye, activated for easy and reliable labeling of antibodies, proteins and other molecules for use as fluorescent probes.

Fluorescein dyes are used in wide-ranging applications including fluorescence microscopy, flow cytometry and immunofluorescence-based assays such as Western blotting and ELISA. FITC and NHS-fluorescein are reactive toward primary amine groups on proteins, peptides and other biomolecules, while fluorescein-5-maleimide and 5-IAF react with free sulfhydryls on cysteine residues.Most activated fluorescein derivatives are mixture of isomers with reactive groups attached at the 5- and 6-positions of the bottom ring. The properties of these isomers are indistinguishable in terms of excitation and emission spectra and for protein applications there is no need to isolate a specific isomer. The single isomer is used in high resolution separation techniques such as sequencing and capillary electrophoresis.

Highlights:

  • Amine-specific labeling: the NHS-ester or isothiocyanate efficiently labels antibodies and other purified proteins at primary amines (lysine side chains) Optimized procedure: following the standard protocol results in antibodies with excellent dye:protein ratios for optimum activity and fluorescence Single-use fluors: no need to weigh tiny amounts powder; kits contain single-use vials of reagent Efficient purification: kits include purification resin and easy-to-use spin columns, ensuring rapid and efficient removal of non-reacted dye and excellent protein recovery
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 g
Product Type NHS-Fluorescein
Solubility DMF (Dimethylformamide)
Content And Storage Store desiccated at -20°C, protect from moisture
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Label or Dye FITC (Fluorescein)
Label Type Classic Dyes
Product Line Pierce
Emission 518
Excitation 494
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.