missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Soluble in alcohol, chloroform, and acetone. Insoluble in water.
Notes
Protect from heat. Store away from oxidizing agents.
Soluble in alcohol, chloroform, and acetone. Insoluble in water.
Notes
Protect from heat. Store away from oxidizing agents.

Specifications
Specifications
Quantity | 1g |
Merck Index | 14,6496 |
Solubility Information | Soluble in alcohol,chloroform,and acetone. Insoluble in water. |
Formula Weight | 484.39 |
Chemical Name or Material | Nicergoline |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z22H045 | Certificate of Analysis | 24/02/2023 | J63295.03,J63295.MC |
Q19I059 | Certificate of Analysis | 28/04/2022 | J63295.03,J63295.MC |
F30X092 | Certificate of Analysis | 08/12/2021 | J63295.03,J63295.MC |
B04Z010 | Certificate of Analysis | 06/12/2021 | J63295.03,J63295.MC |
L11Y024 | Certificate of Analysis | 06/12/2021 | J63295.03,J63295.MC |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nicergoline, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction