missing translation for 'onlineSavingsMsg'
Learn More

Nickel 2-methoxyethoxide, 5% w/v in 2-methoxyethanol, Thermo Scientific™

Product Code. 11374498
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142600-62-2
Molecular Formula C6H14NiO4
Molecular Weight (g/mol) 208.867
MDL Number MFCD01863472
InChI Key KNEAYFOSQJIPKZ-UHFFFAOYSA-N
Synonym nickel 2+ bis 2-methoxyethanolate
PubChem CID 131873637
IUPAC Name 2-methoxyethanolate;nickel(2+)
SMILES COCC[O-].COCC[O-].[Ni+2]

Specifications

Linear Formula Ni(OCH2CH2OCH3)2
Quantity 100 mL
UN Number UN1188
Sensitivity Moisture sensitive
Formula Weight 208.76
Concentration or Composition (by Analyte or Components) 5% w/v in 2-methoxyethanol
Physical Form Liquid
Chemical Name or Material Nickel 2-methoxyethoxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'