missing translation for 'onlineSavingsMsg'
Learn More

Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals

Product Code. 10533951 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
Packaging:
Plastic bottle
Unit Size:
250g
This item is not returnable. View return policy

Product Code. 10533951

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-02-0
Melting Point 1455.0°C
Boiling Point 2732.0°C to 2837.0°C
CAS Min % 99.95
CAS Max % 100.0
Density 8.9
Quantity 250 g
Packaging Plastic bottle
Assay Percent Range 99.97%
Form Sphere
Molecular Formula Ni
MDL Number MFCD00011137 MFCD06798735
Fieser 12,335; 13,197; 17,296
Merck Index 15, 6583
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES [Ni]
IUPAC Name nickel
Molecular Weight (g/mol) 58.69
PubChem CID 935
ChEBI CHEBI:28112
Formula Weight 58.7
Percent Purity 99.97%
Grade Trace Metal Basis
Chemical Name or Material Nickel, p.a.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nickel, spheres, 5 to 11 mm
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H351-Suspected of causing cancer.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.