missing translation for 'onlineSavingsMsg'
Learn More

Nickel-aluminum Alloy, Pure, Powder, for Preparing Catalytic Nickel, Fisher Chemical

Product Code. 10010250 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
Unit Size:
250g
This item is not returnable. View return policy

Product Code. 10010250

Brand: Fisher Chemical N/1460/50

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Gray
CAS 12635-29-9
Melting Point 1350°C
Quantity 250 g
Packaging Plastic powder jar
Form Powder/Solid
Molecular Formula AlNi
MDL Number 167392
Synonym aluminum-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminum nickel
InChI Key NPXOKRUENSOPAO-UHFFFAOYSA-N
SMILES [Al].[Ni]
IUPAC Name aluminum;nickel
Molecular Weight (g/mol) 85.675
PubChem CID 6336846
Formula Weight 85.68g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aluminium-nickel, Raney-type alloy, powder
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H351-Suspected of causing cancer.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.