missing translation for 'onlineSavingsMsg'
Learn More

Nickel arsenide, 99.5% (metals basis excluding Co), Co 0.1-1%, Thermo Scientific™

39.00 € - 148.00 €

Chemical Identifiers

CAS 12068-61-0
Molecular Formula As2Ni
Molecular Weight (g/mol) 208.537
MDL Number MFCD00151450
InChI Key LPZQKJKDQZNVJW-UHFFFAOYSA-N
Synonym 1,2-diarsa-3-nickelacycloprop-1-ene
PubChem CID 91886304
IUPAC Name $l^{2}-arsanylidenearsenic;nickel
SMILES [Ni].[As]=[As]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11333338
View Documents
Thermo Scientific Alfa Aesar
036269.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11343338
View Documents
Thermo Scientific Alfa Aesar
036269.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

12068-61-0
208.537
LPZQKJKDQZNVJW-UHFFFAOYSA-N
91886304
[Ni].[As]=[As]
As2Ni
MFCD00151450
1,2-diarsa-3-nickelacycloprop-1-ene
$l^{2}-arsanylidenearsenic;nickel

Specifications

12068-61-0
As2Ni
UN3077
LPZQKJKDQZNVJW-UHFFFAOYSA-N
$l^{2}-arsanylidenearsenic;nickel
91886304
Odorless
Nickel arsenide
Lump
MFCD00151450
1,2-diarsa-3-nickelacycloprop-1-ene
[Ni].[As]=[As]
208.537
99.5%
(metals basis excluding Co), Co 0.1 to 1%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350i-H372

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 235-103-1

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.