missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Chromium slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.5 in.) length, 99.95% (metals basis), Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Avg. Mol. Wt. or Mol. Wt. Range | Ni:Cr; 80:20 wt% |
Quantity | 50 g |
IUPAC Name | chromium nickel |
Percent Purity | 99.95% |
Odor | Odorless |
Weight | ≈3.4g/slug |
Form | Slug |
Assay | (metals basis) |
Chemical Name or Material | Nickel Chromium slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.5 in.) length |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'