missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chromium, sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 15484805
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Ea.
CAS 11106-97-1
Odor Odorless
Molecular Formula CrNi
MDL Number MFCD00673110
Synonym nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
InChI Key VNNRSPGTAMTISX-UHFFFAOYSA-N
SMILES [Cr].[Ni]
IUPAC Name chromium nickel
Molecular Weight (g/mol) 110.69
PubChem CID 11320920
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Nickel Chromium, sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'