missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chromium wire, 0.143mm (0.0056 in.) dia., Thermo Scientific Chemicals

High resistivity material for electronics industry

34.77 € - 184.00 €

Chemical Identifiers

CAS 11106-97-1
Molecular Formula CrNi
Molecular Weight (g/mol) 110.69
MDL Number MFCD00673110
InChI Key VNNRSPGTAMTISX-UHFFFAOYSA-N
Synonym nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
PubChem CID 11320920
SMILES [Cr].[Ni]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11371479
View Documents Promotion Details
Thermo Scientific Alfa Aesar
013966.H4
100 m
34.77 €
100m
Please to purchase this item. Need a web account? Register with us today!
11381479
View Documents Promotion Details
Thermo Scientific Alfa Aesar
013966.H9
500 m
78.49 €
500m
Please to purchase this item. Need a web account? Register with us today!
11391479
View Documents Promotion Details
Thermo Scientific Alfa Aesar
013966.J5
2000 m This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

High resistivity material for electronics industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

11106-97-1
110.69
VNNRSPGTAMTISX-UHFFFAOYSA-N
11320920
CrNi
MFCD00673110
nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
[Cr].[Ni]

Specifications

11106-97-1
CrNi
nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
[Cr].[Ni]
110.69
Odorless
Wire
Ni:Cr; 80:20 wt%
MFCD00673110
VNNRSPGTAMTISX-UHFFFAOYSA-N
chromium nickel
11320920
≈0.14g/m
Nickel Chromium wire, 0.143mm (0.0056 in.) dia.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H351

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only