missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Copper foil, alloy 400, 0.81mm (0.032 in.) thick, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045169.HB
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel Copper foil, Alloy 400 | |
Ni:Cu; 67:33 wt% | |
300 x 300 mm |
0.81mm (0.032 in.) thick | |
MFCD00801098 | |
Foil |
RUO – Research Use Only