missing translation for 'onlineSavingsMsg'
Learn More

Nickel foil, 0.127mm (0.005in) thick, annealed, 99+% (metals basis), Thermo Scientific Chemicals

Product Code. 11330759
Click to view available options
Quantity:
20 x 30 cm
30 x 100 cm
30 x 500 cm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Quantity 20 x 30 cm
Percent Purity ≥99%
Odor Odorless
Weight ≈67.8g/20x30cm
Form Foil
Assay (metals basis)
Chemical Name or Material Nickel foil, 0.127mm (0.005 in.) thick, Annealed, Temper: soft

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'