missing translation for 'onlineSavingsMsg'
Learn More

Nickel foil, 0.25mm (0.01in) thick, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals

Form: Approx. 2.8g/25 x 50mm

127.64 € - 1885.38 €

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11366388
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012045.FG
25 x 50 mm
127.64 €
Each
Please to purchase this item. Need a web account? Register with us today!
11386388
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012045.FY
50 x 100 mm
518.58 €
Each
Please to purchase this item. Need a web account? Register with us today!
11376388
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012045.HL
100 x 250 mm
1885.38 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-02-0
58.69
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137 MFCD06798735
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
CHEBI:28112
[Ni]

Specifications

Silver to Gray
Silver gray foil
Ni
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
[Ni]
58.69
CHEBI:28112
Foil
7440-02-0
99.994% (Metals basis)
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
935
Odorless
Nickel
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H351-H372

missing translation for 'einecsNumber' : 231-111-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only