missing translation for 'onlineSavingsMsg'
Learn More
Nickel foil, 2.0mm (0.08in) thick, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 11386219

Nickel foil, 2.0mm (0.08in) thick, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11386219
50 x 50 mm
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
This item is not returnable. View return policy

Product Code. 11386219

Brand: Thermo Scientific Alfa Aesar 044825.FI

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Quantity 50 x 50 mm
Percent Purity 99.5%
Odor Odorless
Weight ≈44.5g/50x50mm
Form Foil
Assay (metals basis)
Chemical Name or Material Nickel foil, 2.0mm (0.08 in.) thick

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ26K005 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number044825.HP,044825.FI,044825.GH
Lot NumberM03F053 Certificate TypeCertificate of Analysis Date18/10/2023 Catalog Number044825.HP,044825.FI,044825.GH
Lot NumberQ26I061 Certificate TypeCertificate of Analysis Date24/05/2022 Catalog Number044825.HP,044825.FI,044825.GH
Lot NumberJ28W002 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044825.HP,044825.FI,044825.GH
Lot NumberI02U028 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044825.HP,044825.FI,044825.GH

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel foil, 2.0mm (0.08in) thick, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.