missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) acetate hydrate, 99+%, Thermo Scientific Chemicals

42.91 € - 583.91 €

Chemical Identifiers

CAS 6018-89-9
Molecular Formula C4H14NiO8
Molecular Weight (g/mol) 248.84
MDL Number MFCD00066973
InChI Key OINIXPNQKAZCRL-UHFFFAOYSA-L
Synonym nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate
PubChem CID 62601
IUPAC Name nickel(2+) diacetate tetrahydrate
SMILES O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11366638
View Documents
Thermo Scientific Alfa Aesar
012223.30
250 g
42.91 €
250g
Please to purchase this item. Need a web account? Register with us today!
11356638
View Documents
Thermo Scientific Alfa Aesar
012223.A1
1 kg
128.64 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11346638
View Documents
Thermo Scientific Alfa Aesar
012223.A7
5 kg
583.91 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used in synthesis of polynuclear-nickel polyoxotungstate cluster compounds that are ideal for the design of molecular magnets .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6018-89-9
248.84
OINIXPNQKAZCRL-UHFFFAOYSA-L
62601
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O
C4H14NiO8
MFCD00066973
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate
nickel(2+) diacetate tetrahydrate

Specifications

(decomposition)
White
99+%
MFCD00066973
Soluble in water,alcohol
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O
248.84
176.79 (Anhydrous)
Green crystals
6018-89-9
Crystalline
C4H14NiO8
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate
OINIXPNQKAZCRL-UHFFFAOYSA-L
nickel(2+) diacetate tetrahydrate
62601
Light
Nickel(II) acetate hydrate
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c

H302+H332-H317-H334-H335-H341-H350i-H360D-H372

missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III

missing translation for 'einecsNumber' : 206-761-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Nickel(II) acetate hydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Nickel(II) acetate tetrahydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.