Learn More
Nickel(II) acetate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals
30.90 € - 261.00 €
Chemical Identifiers
CAS | 6018-89-9 |
---|---|
Molecular Formula | C4H14NiO8 |
Molecular Weight (g/mol) | 248.84 |
MDL Number | MFCD00066973 |
InChI Key | OINIXPNQKAZCRL-UHFFFAOYSA-L |
Synonym | nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate |
PubChem CID | 62601 |
SMILES | O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10551354
|
Thermo Scientific Acros
223141000 |
100 g | Plastic bottle |
30.90 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10741881
|
Thermo Scientific Acros
223145000 |
500 g | Plastic bottle |
73.10 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974901
|
Thermo Scientific Acros
223140025 |
2.5 kg | Plastic bottle |
261.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6018-89-9 | |
248.84 | |
OINIXPNQKAZCRL-UHFFFAOYSA-L | |
62601 |
C4H14NiO8 | |
MFCD00066973 | |
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate | |
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O |
Specifications
250.0°C | |
White | |
99% min. (Complexometry) | |
(CH3CO2)2Ni·4H2O | |
14,288; 16,308 | |
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate | |
OINIXPNQKAZCRL-UHFFFAOYSA-L | |
nickel(2+);diacetate;tetrahydrate | |
62601 | |
≥99% | |
Authentic |
6018-89-9 | |
Crystalline Powder, Crystals or Flakes | |
C4H14NiO8 | |
MFCD00066973 | |
14,6498 | |
Solubility in water: 182g/L (20°C). Other solubilities: soluble in alcohol | |
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O | |
248.84 | |
248.86 | |
Analytical | |
Nickel(II) acetate tetrahydrate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective g
GHS Signal Word: Danger
- Nickel(II) acetate tetrahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360D-May damage the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Acetic acid, nickel(2+) salt, Nickel acetate tetrahydrate
RUO – Research Use Only