missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
It is used in electroplating, anodize coatings; textile dyeing mordant; Catalyst. Also, used in synthesis of polynuclear-nickel polyoxotungstate cluster compounds that are ideal for the design of molecular magnets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | (decomposition) |
Color | Green |
Physical Form | Crystalline Aggregates |
Quantity | 25 g |
Linear Formula | Ni(CH3COO)2·4H2O |
UN Number | UN3077 |
Beilstein | 3730764 |
Merck Index | 14,6499 |
Solubility Information | Soluble in water and methanol. Insoluble in diethyl ether,n-octanol. |
Formula Weight | 248.84 (176.79 Anhydrous) |
Mehr anzeigen |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur