missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) bromide, anhydrous, 99%, Thermo Scientific Chemicals

114.57 € - 380.90 €

Chemical Identifiers

CAS 13462-88-9
Molecular Formula Br2Ni
Molecular Weight (g/mol) 218.501
MDL Number MFCD00011141
InChI Key IPLJNQFXJUCRNH-UHFFFAOYSA-L
Synonym nickel bromide, nickel ii bromide, nickel dibromide, nickelous bromide, nickel bromide nibr2, nickel 2+ bromide, unii-41sgh8a2s6, nibr2, nickel ii bromide, anhydrous
PubChem CID 278492
IUPAC Name dibromonickel
SMILES [Ni](Br)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11478650
View Documents
Thermo Scientific Alfa Aesar
053115.18
50 g
114.57 €
50g
Please to purchase this item. Need a web account? Register with us today!
11468650
View Documents
Thermo Scientific Alfa Aesar
053115.30
250 g
380.90 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nickel(II) bromide is used as a source of the bromide ion. It is an important raw material and intermediate used in synthetic chemistry. Further, it is used as a catalyst in cross-coupling reactions.It is used in the preparation of paramagnetic trans-diaquabis(N,N-diethylethylenediamine) nickel(II) bromide and diamagnetic bis(N,N-diethylethylenediamine)nickel(II) bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13462-88-9
218.501
IPLJNQFXJUCRNH-UHFFFAOYSA-L
278492
[Ni](Br)Br
Br2Ni
MFCD00011141
nickel bromide, nickel ii bromide, nickel dibromide, nickelous bromide, nickel bromide nibr2, nickel 2+ bromide, unii-41sgh8a2s6, nibr2, nickel ii bromide, anhydrous
dibromonickel

Specifications

963°C
Crystalline
NiBr2
UN3077
nickel bromide, nickel ii bromide, nickel dibromide, nickelous bromide, nickel bromide nibr2, nickel 2+ bromide, unii-41sgh8a2s6, nibr2, nickel ii bromide, anhydrous
IPLJNQFXJUCRNH-UHFFFAOYSA-L
dibromonickel
278492
99%
Hygroscopic
Nickel (II) bromide, Anhydrous
13462-88-9
Br2Ni
MFCD00011141
14,6502
Soluble in water,alcohol and ether.
[Ni](Br)Br
218.501
218.53
Odorless
5.098 g/mL at 27°C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P302+P352-P304+P340-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H335-H341-H350i-H360D-H372

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 236-665-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Nickel(II) bromide anhydrous

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : Nickel bromide (NiBr2)

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only