Learn More
Nickel(II) carbonate, anhydrous, 98%, Thermo Scientific Chemicals
44.60 € - 151.00 €
Chemical Identifiers
CAS | 3333-67-3 |
---|---|
Molecular Formula | CNiO3 |
Molecular Weight (g/mol) | 118.70 |
MDL Number | MFCD00011144 |
InChI Key | ZULUUIKRFGGGTL-UHFFFAOYSA-L |
Synonym | nickel carbonate, nickelous carbonate, nickel ii carbonate, nickel monocarbonate, unii-wm50squ829, carbonic acid, nickel 2+ salt 1:1, nickel 2+ carbonate nico3, nickel 2+ ion carbonate, nickel carbonate nico3 |
PubChem CID | 18746 |
IUPAC Name | nickel(2+);carbonate |
SMILES | [Ni++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11308127
|
Thermo Scientific Alfa Aesar
022897.18 |
50 g |
44.60 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11318127
|
Thermo Scientific Alfa Aesar
022897.30 |
250 g |
151.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nickel(II) carbonate is used in sulphamate baths, metal phosphating, electroplating and ceramic applications. It acts as a precursor to catalysts and an intermediate in the hydrometallurgical purification of nickel from its ores.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3333-67-3 | |
118.70 | |
ZULUUIKRFGGGTL-UHFFFAOYSA-L | |
18746 | |
[Ni++].[O-]C([O-])=O |
CNiO3 | |
MFCD00011144 | |
nickel carbonate, nickelous carbonate, nickel ii carbonate, nickel monocarbonate, unii-wm50squ829, carbonic acid, nickel 2+ salt 1:1, nickel 2+ carbonate nico3, nickel 2+ ion carbonate, nickel carbonate nico3 | |
nickel(2+);carbonate |
Specifications
3333-67-3 | |
Powder | |
CNiO3 | |
nickel carbonate, nickelous carbonate, nickel ii carbonate, nickel monocarbonate, unii-wm50squ829, carbonic acid, nickel 2+ salt 1:1, nickel 2+ carbonate nico3, nickel 2+ ion carbonate, nickel carbonate nico3 | |
ZULUUIKRFGGGTL-UHFFFAOYSA-L | |
nickel(2+);carbonate | |
18746 | |
Light powder |
Beige | |
98% | |
MFCD00011144 | |
Soluble in dilute acids. Insoluble in cold water. | |
[Ni++].[O-]C([O-])=O | |
118.70 | |
118.72 | |
Nickel(II) carbonate, anhydrous |
Safety and Handling
Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
H302+H332-H315-H317-H319-H334-H341-H350i-H360D-H372
missing translation for 'dotInformation' : Hazard Class: 9, Packing Group: III
missing translation for 'einecsNumber' : 222-068-2
missing translation for 'rtecsNumber' : QR6200000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only