missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) carbonate (basic) hydrate, Ni 40% min, typically 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11369237
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Nickel Plating pH Adjuster, Catalysts, Ceramic Colors and Glazes

Nickel plating pH adjuster, catalysts, ceramic colors and glazes.Nickel(II) carbonate (basic) hydrate is used as a precursor to catalysts and in ceramic applications. It acts as an intermediate in the hydrometallurgical purification of nickel from its ores. It is also used in electroplating of nickel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155775-31-8
Molecular Formula CH4Ni3O7
Molecular Weight (g/mol) 304.12
MDL Number MFCD00016262
InChI Key ATYNVYNPWGEMMI-UHFFFAOYSA-H
Synonym nickel 3+ ;carbonate;hydroxide;hydrate, nickel carbonate, basic hydrate trace metals basis
PubChem CID 86176052
IUPAC Name trinickel(2+) tetrahydroxide carbonate
SMILES [OH-].[OH-].[OH-].[OH-].[Ni++].[Ni++].[Ni++].[O-]C([O-])=O

Specifications

Color Green
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 304.18 (Anhydrous)
Odor Odorless
Appearance Green powder
Chemical Name or Material Nickel(II) carbonate (basic) hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.