Learn More
Nickel(II) carbonate (basic) hydrate, Puratronic™, 99.996% (metals basis), Thermo Scientific™
Chemical Identifiers
CAS | 155775-31-8 |
---|---|
Molecular Formula | CH4Ni3O7 |
Molecular Weight (g/mol) | 304.12 |
MDL Number | MFCD00016262 |
InChI Key | ATYNVYNPWGEMMI-UHFFFAOYSA-H |
Synonym | nickel 3+ ;carbonate;hydroxide;hydrate, nickel carbonate, basic hydrate trace metals basis |
PubChem CID | 86176052 |
IUPAC Name | nickel(3+);carbonate;hydroxide;hydrate |
SMILES | [OH-].[OH-].[OH-].[OH-].[Ni++].[Ni++].[Ni++].[O-]C([O-])=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
155775-31-8 | |
304.12 | |
ATYNVYNPWGEMMI-UHFFFAOYSA-H | |
86176052 | |
[OH-].[OH-].[OH-].[OH-].[Ni++].[Ni++].[Ni++].[O-]C([O-])=O |
CH4Ni3O7 | |
MFCD00016262 | |
nickel 3+ ;carbonate;hydroxide;hydrate, nickel carbonate, basic hydrate trace metals basis | |
nickel(3+);carbonate;hydroxide;hydrate |
Specifications
155775-31-8 | |
Powder | |
NiCO3·2Ni(OH)2·xH2O | |
UN3077 | |
Insoluble in water. | |
[OH-].[OH-].[OH-].[OH-].[Ni++].[Ni++].[Ni++].[O-]C([O-])=O | |
304.12 | |
304.18 (Anhydrous) | |
Puratronic™ | |
Nickel(II) carbonate (basic) hydrate |
Green | |
CH4Ni3O7 | |
MFCD00016262 | |
nickel 3+ ;carbonate;hydroxide;hydrate, nickel carbonate, basic hydrate trace metals basis | |
ATYNVYNPWGEMMI-UHFFFAOYSA-H | |
nickel(3+);carbonate;hydroxide;hydrate | |
86176052 | |
99.996% | |
(metals basis) |
Safety and Handling
GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
H302+H332-H315-H317-H319-H334-H341-H350i-H360D-H372
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 240-408-8
missing translation for 'rtecsNumber' : QR6260000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures