Learn More
Nickel(II) chloride, anhydrous, 98%, Thermo Scientific Chemicals
Description
Nickel(II) chloride is a mild Lewis acid used for the regioselective isomerization of dienols and selective reduction of alkenes to alkanes in presence of Lithium aluminum hydride. It is used as a precursor to nickel boride, finely divided nickel. It acts as a catalyst to prepare dialkyl arylphosphonates from phosphites and aryl iodide. It finds application in the electroplating of nickel onto other metal items. It is also used in the synthesis of semiconducting metal-containing polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Nickel(II) chloride is a mild Lewis acid used for the regioselective isomerization of dienols and selective reduction of alkenes to alkanes in presence of Lithium aluminum hydride. It is used as a precursor to nickel boride, finely divided nickel. It acts as a catalyst to prepare dialkyl arylphosphonates from phosphites and aryl iodide. It finds application in the electroplating of nickel onto other metal items. It is also used in the synthesis of semiconducting metal-containing polymers.
Solubility
Soluble in ethylene glycol, ethanol, hydrazine and ammonium hydroxide. Slightly soluble in water. Insoluble in nitric acid.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with peroxides.
Specifications
Specifications
Melting Point | 1,001°C |
Quantity | 25 g |
Linear Formula | NiCl2 |
UN Number | UN3288 |
Merck Index | 14,6505 |
Solubility Information | Soluble in ethylene glycol,ethanol,hydrazine and ammonium hydroxide. Slightly soluble in water. Insoluble in nitric acid. |
Formula Weight | 129.6 |
Percent Purity | 98% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
- Nickel(II) chloride, anhydrous
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H331-Toxic if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360D-May damage the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Nickel(II) chloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.